Gene Transcripts

Gene Symbol
MNS1
Entrez Gene ID
55329
Gene Name
meiosis specific nuclear structural 1
Gene Aliases
SPATA40
Chromosome Location
Chr. 15: 56428731 - 56465137 on Build GRCh38
UniGene ID
Hs.444483
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018365.2 NP_060835.1 9 1535
GenBank mRNA AK002084.1 - 9 1535
AK057542.1 - 8 1335
BC031046.1 - 9 1448 5273227
BC034991.2 - 9 1502 4823345

View Full Product Details