Gene Transcripts

Gene Symbol
MYO1H
Entrez Gene ID
283446
Gene Name
myosin IH
Gene Aliases
Chromosome Location
Chr. 12: 109388277 - 109448371 on Build GRCh38
UniGene ID
Hs.512008
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001101421.3 NP_001094891.3 31 3961
GenBank mRNA AK094906.1 - 7 1925

View Full Product Details