Gene Transcripts

Gene Symbol
CNP
Entrez Gene ID
1267
Gene Name
2',3'-cyclic nucleotide 3' phosphodiesterase
Gene Aliases
CNP1
Chromosome Location
Chr. 17: 41966741 - 41977736 on Build GRCh38
UniGene ID
Hs.273621
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033133.4 NP_149124.3 3 950
XM_006721701.3 XP_006721764.1 3 974
XM_011524340.2 XP_011522642.1 3 935
GenBank mRNA AK026876.1 - 3 907
AK097833.1 - 3 798
AK098384.1 - 3 896
AK294179.1 - 2 223
AK295351.1 - 4 531
BC001362.2 - 3 887 3051112
BC006392.2 - 3 909 4126696
BC011046.1 - 3 871 4179083
BC028040.1 - 3 911 5248370
M19650.1 - 2 759

View Full Product Details