Gene Transcripts
- Gene Symbol
- LSM14A
- Entrez Gene ID
- 26065
- Gene Name
- LSM14A, mRNA processing body assembly factor
- Gene Aliases
- C19orf13, FAM61A, RAP55, RAP55A
- Chromosome Location
- Chr. 19: 34172428 - 34229515 on Build GRCh38
- UniGene ID
- Hs.744009
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001114093.1 |
NP_001107565.1
|
2, 3 |
478 |
|
NM_015578.2 |
NP_056393.2
|
2, 3 |
478 |
|
XM_005258719.4 |
XP_005258776.1
|
2, 3 |
497 |
|
XM_005258720.4 |
XP_005258777.1
|
2, 3 |
497 |
|
XM_005258721.4 |
XP_005258778.1
|
2, 3 |
497 |
|
XM_011526708.2 |
XP_011525010.1
|
2, 3 |
497 |
|
XM_017026575.1 |
XP_016882064.1
|
2, 3 |
497 |
|
XM_017026576.1 |
XP_016882065.1
|
2, 3 |
497 |
|
XM_017026577.1 |
XP_016882066.1
|
2, 3 |
497 |
|
GenBank mRNA |
AB069974.1 |
- |
2, 3 |
357 |
|
AB069975.1 |
- |
2, 3 |
357 |
|
AK027369.1 |
- |
1, 2 |
88 |
|
AK027643.1 |
- |
2, 3 |
359 |
|
AK300208.1 |
- |
2, 3 |
400 |
|
AL834398.1 |
- |
2, 3 |
387 |
|
BC016842.1 |
- |
2, 3 |
478 |
3902704 |
View Full Product Details