Gene Transcripts

Gene Symbol
NDUFS5
Entrez Gene ID
4725
Gene Name
NADH:ubiquinone oxidoreductase subunit S5
Gene Aliases
CI-15k, CI15K
Chromosome Location
Chr. 1: 39026295 - 39034636 on Build GRCh38
UniGene ID
Hs.632385
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF020352.1 - 493
AF047434.1 - 504
AF086268.1 - 494
BC001884.1 - 461 3544747

View Full Product Details