Gene Transcripts

Gene Symbol
Cables2
Entrez Gene ID
252966
Gene Name
CDK5 and Abl enzyme substrate 2
Gene Aliases
ik3-2
Chromosome Location
Chr. 2: 180258539 - 180273490 on Build GRCm38
UniGene ID
Mm.386884
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145851.2 NP_665850.2 10 1407
GenBank mRNA AK050121.1 - 7 3029
BC024565.1 - 3 418
BC036954.1 - 5 798
BC037028.1 - 5 798
BC052789.1 - 4 749

View Full Product Details