Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008550.2 | NP_032576.2 | 13 | 2344 | |
GenBank mRNA | AB006458.1 | - | 13 | 2322 | |
AK004900.1 | - | 13 | 2298 | ||
AK040388.1 | - | 13 | 2305 | ||
AK075649.1 | - | 5 | 1011 | ||
AK165376.1 | - | 13 | 2337 | ||
BC066211.1 | - | 13 | 2295 | 30101816 |