Gene Transcripts
- Gene Symbol
- Nr3c1
- Entrez Gene ID
- 14815
- Gene Name
- nuclear receptor subfamily 3, group C, member 1
- Gene Aliases
- GR, Grl-1, Grl1
- Chromosome Location
- Chr. 18: 39410545 - 39519416 on Build GRCm38
- UniGene ID
- Mm.129481
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_008173.3 |
NP_032199.3
|
2, 3 |
1420 |
|
GenBank mRNA |
AK037733.1 |
- |
2, 3 |
1300 |
|
AK155200.1 |
- |
4, 5 |
1502 |
|
AK156323.1 |
- |
4, 5 |
1496 |
|
BC016462.1 |
- |
4, 5 |
1503 |
|
BC129912.1 |
- |
3, 4 |
1398 |
40111802 |
BC129913.1 |
- |
3, 4 |
1398 |
40111803 |
DQ504162.1 |
- |
2, 3 |
1404 |
|
HM236293.1 |
- |
3, 4 |
1380 |
|
X04435.1 |
- |
3, 4 |
1398 |
|
X13358.1 |
- |
3, 4 |
1380 |
|
View Full Product Details