Gene Transcripts

Gene Symbol
Mlst8
Entrez Gene ID
56716
Gene Name
MTOR associated protein, LST8 homolog (S. cerevisiae)
Gene Aliases
0610033N12Rik, AA409454, AI505104, AI851821, Gbl
Chromosome Location
Chr. 17: 24473550 - 24479114 on Build GRCm38
UniGene ID
Mm.289516
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001252463.1 NP_001239392.1 5 494
NM_001252464.1 NP_001239393.1 5 469
NM_001252465.1 NP_001239394.1 3 278
NM_019988.5 NP_064372.2 5 489
GenBank mRNA AK002751.1 - 5 455
AK038515.1 - 4 987
AK040100.1 - 5 490
AK048102.1 - 5 453
AK077680.1 - 5 460
AK083731.1 - 5 430
AK088951.1 - 5 463
BC015279.1 - 5 448 4222934
BM947104.1 - 3 260
CF750475.1 - 5 458

View Full Product Details