Gene Transcripts

Gene Symbol
Prss27
Entrez Gene ID
213171
Gene Name
protease, serine 27
Gene Aliases
CAPH2, Mpn, marapsin
Chromosome Location
Chr. 17: 24038243 - 24048378 on Build GRCm38
UniGene ID
Mm.133083
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175440.4 NP_780649.1 6 1089
GenBank mRNA AK080281.1 - 6 1090
BC117009.1 - 6 1072 40126328
BC117011.1 - 6 1072 40126330

View Full Product Details