Gene Transcripts

Gene Symbol
Mslnl
Entrez Gene ID
328783
Gene Name
mesothelin-like
Gene Aliases
4732467B22
Chromosome Location
Chr. 17: 25733503 - 25748530 on Build GRCm38
UniGene ID
Mm.183275
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177822.3 NP_808490.1 10, 11 1429
GenBank mRNA AK028891.1 - 10, 11 1429
BC052484.1 - 9, 10 1165 6312790

View Full Product Details