Gene Transcripts

Gene Symbol
Mfsd5
Entrez Gene ID
106073
Gene Name
major facilitator superfamily domain containing 5
Gene Aliases
AA409375, AW556797, D15Mgi27
Chromosome Location
Chr. 15: 102279456 - 102281744 on Build GRCm38
UniGene ID
Mm.287740
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134100.4 NP_598861.1 2 378
GenBank mRNA AK087550.1 - 2 381
AK089268.1 - 2 381
AK165774.1 - 2 350
AK197827.1 - 1 198
BC009140.1 - 1 285 3598007

View Full Product Details