Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_134163.4 | NP_598924.1 | 5 | 689 | |
GenBank mRNA | AB077700.1 | - | 4 | 595 | |
AF466292.1 | - | 4 | 595 | ||
AJ427920.1 | - | 4 | 595 | ||
AK040522.1 | - | 4 | 443 | ||
AK050908.1 | - | 4 | 408 | ||
AK088971.1 | - | 5 | 702 | ||
AK167342.1 | - | 4 | 408 | ||
BC057157.1 | - | 5 | 927 | 5348959 |