Gene Transcripts
- Gene Symbol
- NAP1L4
- Entrez Gene ID
- 4676
- Gene Name
- nucleosome assembly protein 1 like 4
- Gene Aliases
- NAP1L4b, NAP2, NAP2L, hNAP2
- Chromosome Location
- Chr. 11: 2944430 - 2992377 on Build GRCh38
- UniGene ID
- Hs.731784
- Species
- Homo sapiens
- Gene Symbol
- CD58
- Entrez Gene ID
- 965
- Gene Name
- CD58 molecule
- Gene Aliases
- LFA-3, LFA3, ag3
- Chromosome Location
- Chr. 1: 116487140 - 116571093 on Build GRCh38
- UniGene ID
- Hs.34341
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
EF491785.1 |
- |
2 |
179 |
|
View Full Product Details