Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001317182.1 | NP_001304111.1 | 2 | 304 | |
NM_001317183.1 | NP_001304112.1 | 2 | 301 | ||
NM_006838.3 | NP_006829.1 | 2 | 304 | ||
NR_133673.1 | 2 | 304 | |||
XM_005268583.3 | XP_005268640.1 | 2 | 223 | ||
GenBank mRNA | AK091730.1 | - | 2 | 177 | |
AK125296.1 | - | 2 | 830 | ||
AK300836.1 | - | 2 | 269 | ||
AK315559.1 | - | 2 | 241 | ||
BC013782.1 | - | 2 | 182 | 4340717 | |
U13261.1 | - | 2 | 204 |