Gene Transcripts

Gene Symbol
Zap70
Entrez Gene ID
22637
Gene Name
zeta-chain (TCR) associated protein kinase
Gene Aliases
Srk, ZAP-70, mrtle, mur
Chromosome Location
Chr. 1: 36761798 - 36782821 on Build GRCm38
UniGene ID
Mm.8038
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289765.1 NP_001276694.1 4 881
NM_001289766.1 NP_001276695.1 4 846
NM_009539.3 NP_033565.2 4 838
GenBank mRNA AK039883.1 - 4 775
AK089056.1 - 4 811
BC029727.1 - 4 796 4925739
U04379.1 - 4 792
U77667.1 - 3 684

View Full Product Details