Gene Transcripts

Gene Symbol
Tpbg
Entrez Gene ID
21983
Gene Name
trophoblast glycoprotein
Gene Aliases
5T4, AW495680, WAIF1
Chromosome Location
Chr. 9: 85842380 - 85847055 on Build GRCm38
UniGene ID
Mm.20864
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164792.1 NP_001158264.1 2 1651
NM_011627.4 NP_035757.2 2 1760
GenBank mRNA AK049377.1 - 1 103
AK051162.1 - 2 1731
AK143519.1 - 2 1590
AK165553.1 - 2 1646
AK165597.1 - 2 1763
BC058198.1 - 2 1619 5353871

View Full Product Details