Gene Transcripts

Gene Symbol
Mmp17
Entrez Gene ID
23948
Gene Name
matrix metallopeptidase 17
Gene Aliases
MT4-MMP, MT4MMP, MTMMP4
Chromosome Location
Chr. 5: 129584174 - 129608211 on Build GRCm38
UniGene ID
Mm.42047
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011846.4 NP_035976.3 2, 3 362
GenBank mRNA AB021224.2 - 2, 3 368
AJ010731.1 - 2, 3 366
AK079221.1 - 133
AK143521.1 - 2, 3 361
BC051917.1 - 2, 3 362 5716458

View Full Product Details