Gene Transcripts

Gene Symbol
Nos1ap
Entrez Gene ID
192363
Gene Name
nitric oxide synthase 1 (neuronal) adaptor protein
Gene Aliases
Capon
Chromosome Location
Chr. 13: 88653123 - 88943976 on Build Rnor_6.0
UniGene ID
Rn.9903
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138922.1 NP_620277.1 1 481
GenBank mRNA AF037071.1 - 1 481

View Full Product Details