Gene Transcripts

Gene Symbol
Egf
Entrez Gene ID
25313
Gene Name
epidermal growth factor
Gene Aliases
Chromosome Location
Chr. 2: 235049350 - 235062491 on Build Rnor_6.0
UniGene ID
Rn.6075
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012842.1 NP_036974.1 597
GenBank mRNA BC062030.1 - 582
JX149564.1 - 441
JX437943.1 - 441
U04842.1 - 597

Gene Symbol
LOC100910178
Entrez Gene ID
100910178
Gene Name
pro-epidermal growth factor-like
Gene Aliases
Chromosome Location
Chr. 2: 68820605 - 68895943 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008760814.1 XP_008759036.1 2 608
XM_008760815.1 XP_008759037.1 2 607
XM_008760816.1 XP_008759038.1 2 368

View Full Product Details