Gene Transcripts

Gene Symbol
FAM53B
Entrez Gene ID
9679
Gene Name
family with sequence similarity 53 member B
Gene Aliases
KIAA0140, bA12J10.2, smp
Chromosome Location
Chr. 10: 124619294 - 124744361 on Build GRCh38
UniGene ID
Hs.129195
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014661.3 NP_055476.3 4 692
GenBank mRNA AK127343.1 - 4 886
BC031654.1 - 4 600 5174078
BX344666.2 - 4 690
D50930.1 - 4 381

View Full Product Details