Gene Transcripts

Gene Symbol
Pyroxd1
Entrez Gene ID
297708
Gene Name
pyridine nucleotide-disulphide oxidoreductase domain 1
Gene Aliases
Chromosome Location
Chr. 4: 176565733 - 176581971 on Build Rnor_6.0
UniGene ID
Rn.154505
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004234.1 NP_001004234.1 7 701
GenBank mRNA BC079377.1 - 7 701

View Full Product Details