Gene Transcripts

Gene Symbol
Lca5l
Entrez Gene ID
385668
Gene Name
Leber congenital amaurosis 5-like
Gene Aliases
4921526F01Rik, Lcca5l
Chromosome Location
Chr. 16: 96158405 - 96193993 on Build GRCm38
UniGene ID
Mm.24626
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001492.2 NP_001001492.2 11 2808
GenBank mRNA AK029554.1 - 11 2803
BC141285.1 - 10 2655 9055897
BC145611.1 - 10 2654 9054177

View Full Product Details