Gene Transcripts

Gene Symbol
Adi1
Entrez Gene ID
298934
Gene Name
acireductone dioxygenase 1
Gene Aliases
Alp1, Fe-ARD, MTCBP-1, Ni-ARD
Chromosome Location
Chr. 6: 47940211 - 47947173 on Build Rnor_6.0
UniGene ID
Rn.12694
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199097.1 NP_954528.1 4 923
GenBank mRNA AY346335.1 - 4 923
BC092562.1 - 4 918 7326245

View Full Product Details