Gene Transcripts

Gene Symbol
Lhfpl5
Entrez Gene ID
328789
Gene Name
lipoma HMGIC fusion partner-like 5
Gene Aliases
9330179O15Rik, Tmhs, hscy
Chromosome Location
Chr. 17: 28575371 - 28583593 on Build GRCm38
UniGene ID
Mm.284760
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026571.2 NP_080847.2 3, 4 970
GenBank mRNA AK020389.1 - 3, 4 976
AK020670.1 - 3, 4 958
BC099483.1 - 3, 4 868 30848018

View Full Product Details