Gene Transcripts

Gene Symbol
Olr1006
Entrez Gene ID
288886
Gene Name
olfactory receptor 1006
Gene Aliases
ratchr7-6593123-6592173_ORF
Chromosome Location
Chr. 7: 6096690 - 6097640 on Build Rnor_6.0
UniGene ID
Rn.143366
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000075.1 NP_001000075.1 1 480

View Full Product Details