Gene Transcripts

Gene Symbol
Mrgprb4
Entrez Gene ID
233230
Gene Name
MAS-related GPR, member B4
Gene Aliases
MrgB4
Chromosome Location
Chr. 7: 48198213 - 48199178 on Build GRCm38
UniGene ID
Mm.377250
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_205795.1 NP_991364.1 1 736
GenBank mRNA BC107202.1 - 1 729
BC107203.1 - 1 729

View Full Product Details