Gene Transcripts

Gene Symbol
Mrgprg
Entrez Gene ID
309133
Gene Name
MAS-related GPR, member G
Gene Aliases
Mrgg
Chromosome Location
Chr. 1: 216952942 - 216955673 on Build Rnor_6.0
UniGene ID
Rn.112764
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_203470.1 NP_982296.1 1 367
XM_008760116.1 XP_008758338.1 2 591
XM_008760117.1 XP_008758339.1 2 473

View Full Product Details