Gene Transcripts

Gene Symbol
NYX
Entrez Gene ID
60506
Gene Name
nyctalopin
Gene Aliases
CLRP, CSNB1, CSNB1A, CSNB4, NBM1
Chromosome Location
Chr. X: 41447460 - 41476414 on Build GRCh38
UniGene ID
Hs.302019
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022567.2 NP_072089.1 2 1993
XM_017029709.1 XP_016885198.1 3 1661
GenBank mRNA AF254868.1 - 3 1661
AJ278865.1 - 2 2019

View Full Product Details