Gene Transcripts

Gene Symbol
NKD1
Entrez Gene ID
85407
Gene Name
naked cuticle homolog 1
Gene Aliases
Naked1
Chromosome Location
Chr. 16: 50548330 - 50649249 on Build GRCh38
UniGene ID
Hs.187578
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033119.4 NP_149110.1 9 996
XM_011523399.2 XP_011521701.1 12 1307
XM_011523400.2 XP_011521702.1 7 598
XM_017023804.1 XP_016879293.1 11 2526
GenBank mRNA AB062886.1 - 9 777
AF289584.1 - 9 1048
AF358135.1 - 9 779
AK314930.1 - 9 930
AY061883.1 - 9 930
BC051288.1 - 9 996 6646089

View Full Product Details