Gene Transcripts
- Gene Symbol
- LRRC39
- Entrez Gene ID
- 127495
- Gene Name
- leucine rich repeat containing 39
- Gene Aliases
- Chromosome Location
- Chr. 1: 100148448 - 100178273 on Build GRCh38
- UniGene ID
- Hs.44277
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001256385.1 |
NP_001243314.1
|
3, 4 |
381 |
|
NM_001256386.1 |
NP_001243315.1
|
3, 4 |
381 |
|
NM_001256387.1 |
NP_001243316.1
|
2, 3 |
324 |
|
NM_144620.3 |
NP_653221.1
|
3, 4 |
364 |
|
GenBank mRNA |
AK096892.1 |
- |
3, 4 |
307 |
|
AL711357.1 |
- |
3, 4 |
381 |
|
AL833417.1 |
- |
3, 4 |
329 |
|
AW961910.1 |
- |
3, 4 |
354 |
|
AY358131.1 |
- |
2, 3 |
297 |
|
BC009613.1 |
- |
3, 4 |
306 |
4245944 |
View Full Product Details