Gene Transcripts

Gene Symbol
MORC2-AS1
Entrez Gene ID
150291
Gene Name
MORC2 antisense RNA 1
Gene Aliases
C22orf27, NCRNA00325
Chromosome Location
Chr. 22: 30922308 - 30926654 on Build GRCh38
UniGene ID
Hs.592202
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026920.1 2 859
GenBank mRNA AK093120.1 - 2 758
AK098685.1 - 1 240
BC042980.1 - 3 998
BC065553.1 - 2 752
BC117160.1 - 1 494
BC117186.1 - 1 494

Gene Symbol
MORC2
Entrez Gene ID
22880
Gene Name
MORC family CW-type zinc finger 2
Gene Aliases
CMT2Z, ZCW3, ZCWCC1
Chromosome Location
Chr. 22: 30925130 - 30968497 on Build GRCh38
UniGene ID
Hs.555918
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BM683680.1 - 1 266

View Full Product Details