Gene Transcripts

Gene Symbol
ENPP6
Entrez Gene ID
133121
Gene Name
ectonucleotide pyrophosphatase/phosphodiesterase 6
Gene Aliases
NPP6
Chromosome Location
Chr. 4: 184088706 - 184217961 on Build GRCh38
UniGene ID
Hs.297814
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153343.3 NP_699174.1 2 545
GenBank mRNA AK057370.1 - 2 490
AY358676.1 - 2 486
BC035035.2 - 2 450 5259487

View Full Product Details