Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001286075.1 | NP_001273004.1 | 2 | 82 | |
NM_004320.4 | NP_004311.1 | 4 | 433 | ||
NM_173201.3 | NP_775293.1 | 4 | 433 | ||
GenBank mRNA | AK128456.1 | - | 2 | 72 | |
AK291314.1 | - | 4 | 362 | ||
AL706356.1 | - | 2 | 82 | ||
BC037354.1 | - | 4 | 361 | ||
BX537389.1 | - | 4 | 429 | ||
BX537784.1 | - | 4 | 431 | ||
BX647365.1 | - | 4 | 429 | ||
BX647366.1 | - | 4 | 425 | ||
BX647367.1 | - | 4 | 433 | ||
BX648544.1 | - | 4 | 430 | ||
BX649072.1 | - | 4 | 422 |