Gene Transcripts
- Gene Symbol
- Obp2b
- Entrez Gene ID
- 286933
- Gene Name
- odorant binding protein 2B
- Gene Aliases
- Obp2a, Obp2aa, Ry2g12
- Chromosome Location
- Chr. 3: 3167744 - 3181159 on Build Rnor_6.0
- UniGene ID
- Rn.11371
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_173148.1 |
NP_775171.1
|
|
359 |
|
GenBank mRNA |
M76734.1 |
- |
|
359 |
|
View Full Product Details