Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001033409.3 | NP_001028581.1 | 15 | 1531 | |
GenBank mRNA | AK052873.1 | - | 10 | 731 | |
AK085901.1 | - | 15 | 1490 | ||
AK137380.1 | - | 15 | 1532 | ||
BC141210.1 | - | 15 | 1483 | 9055822 |