Gene Transcripts
- Gene Symbol
- Ly6c
- Entrez Gene ID
- 56778
- Gene Name
- Ly6-C antigen
- Gene Aliases
- Chromosome Location
- Chr. 7|NW_007906224: 20092 - 23957 on Build Rnor_6.0
- UniGene ID
- Rn.40119
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_020103.1 |
NP_064488.1
|
2 |
83 |
|
GenBank mRNA |
M30691.1 |
- |
2 |
81 |
|
View Full Product Details