Gene Transcripts

Gene Symbol
F2
Entrez Gene ID
29251
Gene Name
coagulation factor II
Gene Aliases
Chromosome Location
Chr. 3: 80529468 - 80542993 on Build Rnor_6.0
UniGene ID
Rn.54498
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022924.2 NP_075213.2 4 307
GenBank mRNA EX492416.1 - 4 328
X52835.1 - 4 372

View Full Product Details