Gene Transcripts

Gene Symbol
Cubn
Entrez Gene ID
80848
Gene Name
cubilin (intrinsic factor-cobalamin receptor)
Gene Aliases
GP280, IFCR, MGA1
Chromosome Location
Chr. 17: 80584921 - 80807181 on Build Rnor_6.0
UniGene ID
Rn.3236
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053332.2 NP_445784.1 2 140
GenBank mRNA AF022247.1 - 2 140

View Full Product Details