Gene Transcripts

Gene Symbol
Nkg7
Entrez Gene ID
171062
Gene Name
natural killer cell granule protein 7
Gene Aliases
Chromosome Location
Chr. 1: 98492873 - 98493940 on Build Rnor_6.0
UniGene ID
Rn.20448
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133540.1 NP_598224.1 3 524
GenBank mRNA AF082535.1 - 3 524
FQ222255.1 - 3 530
FQ226323.1 - 3 530

View Full Product Details