Gene Transcripts

Gene Symbol
Mrc2
Entrez Gene ID
17534
Gene Name
mannose receptor, C type 2
Gene Aliases
Endo180, mKIAA0709, uPARAP
Chromosome Location
Chr. 11: 105292576 - 105351146 on Build GRCm38
UniGene ID
Mm.235616
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008626.3 NP_032652.3 3 683
GenBank mRNA AK028157.1 - 3 674
BC116642.1 - 3 609 40086489
U56734.1 - 3 646

View Full Product Details