Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008611.4 | NP_032637.3 | 4 | 689 | |
GenBank mRNA | AK089234.1 | - | 4 | 660 | |
AK089336.1 | - | 4 | 664 | ||
AK089381.1 | - | 4 | 661 | ||
AK089516.1 | - | 4 | 650 | ||
AK137468.1 | - | 4 | 661 | ||
AK154937.1 | - | 4 | 660 | ||
AK155740.1 | - | 4 | 660 | ||
AK155832.1 | - | 4 | 660 | ||
AK155901.1 | - | 4 | 663 | ||
AK170177.1 | - | 4 | 660 | ||
AK170388.1 | - | 4 | 662 | ||
AK170760.1 | - | 4 | 660 | ||
AK171168.1 | - | 4 | 660 | ||
AK171613.1 | - | 4 | 662 | ||
BC042742.1 | - | 4 | 670 | 5032770 | |
U96696.1 | - | 4 | 655 | ||
Y13342.1 | - | 4 | 587 |