Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013933.3 | NP_038961.2 | 5 | 783 | |
GenBank mRNA | AF115503.1 | - | 5 | 505 | |
AF157497.1 | - | 5 | 724 | ||
AK003576.1 | - | 5 | 724 | ||
AK012357.1 | - | 5 | 538 | ||
AK149040.1 | - | 390 | |||
AK159582.1 | - | 5 | 725 | ||
AK167381.1 | - | 5 | 784 | ||
AK168824.1 | - | 5 | 730 | ||
BC003866.1 | - | 5 | 769 | 3490082 |