Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001285833.1 | NP_001272762.1 | 20 | 3621 | |
NM_001285835.1 | NP_001272764.1 | 18 | 3504 | ||
NM_015760.5 | NP_056575.1 | 18 | 3508 | ||
GenBank mRNA | AB042745.1 | - | 18 | 3359 | |
AF261944.1 | - | 18 | 3364 | ||
AK149398.1 | - | 18 | 3358 | ||
AK149492.1 | - | 18 | 3359 |