Gene Transcripts

Gene Symbol
Olfr690
Entrez Gene ID
56860
Gene Name
olfactory receptor 690
Gene Aliases
MOR31-2, Ors18
Chromosome Location
Chr. 7: 105329217 - 105330278 on Build GRCm38
UniGene ID
Mm.377127
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020290.2 NP_064686.2 1 771
GenBank mRNA BC137757.1 - 1 771

View Full Product Details