Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025571.1 | NP_079847.1 | 3, 4 | 314 | |
GenBank mRNA | AB073618.1 | - | 3, 4 | 287 | |
AF349454.1 | - | 3, 4 | 299 | ||
AK003227.1 | - | 3, 4 | 304 | ||
AK008380.1 | - | 3, 4 | 314 | ||
BC024346.1 | - | 3, 4 | 294 | 4989038 | |
BC096419.1 | - | 3, 4 | 303 | 1546951 |