Gene Transcripts
- Gene Symbol
- Rpl15
- Entrez Gene ID
- 66480
- Gene Name
- ribosomal protein L15
- Gene Aliases
- 2510008H07Rik
- Chromosome Location
- Chr. 14: 18267823 - 18271024 on Build GRCm38
- UniGene ID
- Mm.2050
- Species
- Mus musculus
- Gene Symbol
- Gm6937
- Entrez Gene ID
- 628994
- Gene Name
- ribosomal protein L15 pseudogene
- Gene Aliases
- EG628994
- Chromosome Location
- Chr. 19: 42159631 - 42160768 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC089359.1 |
- |
|
771 |
|
View Full Product Details