Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026343.2 | NP_080619.2 | 4 | 353 | |
GenBank mRNA | AK011400.1 | - | 4 | 347 | |
AK014718.1 | - | 3 | 292 | ||
AK018526.1 | - | 4 | 343 | ||
AK034718.1 | - | 4 | 342 | ||
AK135240.1 | - | 4 | 349 | ||
AK154503.1 | - | 4 | 354 | ||
BC028639.1 | - | 4 | 340 | 1243532 |