Gene Transcripts

Gene Symbol
Chic2
Entrez Gene ID
74277
Gene Name
cysteine-rich hydrophobic domain 2
Gene Aliases
1700081B18Rik, 4930502K01Rik, BTL
Chromosome Location
Chr. 5: 75006424 - 75044626 on Build GRCm38
UniGene ID
Mm.239814
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK006962.1 - 3 563
AK172289.1 - 3 556
BC048435.1 - 4 316
BC116971.1 - 3 373 40126290
BC116973.1 - 3 373 40126292

View Full Product Details