Gene Transcripts

Gene Symbol
Gm5795
Entrez Gene ID
544990
Gene Name
predicted gene 5795
Gene Aliases
Chromosome Location
Chr. 14: 3188284 - 3196683 on Build GRCm38
UniGene ID
Mm.360540
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270806.1 NP_001257735.1 1 149
GenBank mRNA BU946301.1 - 1 152

Gene Symbol
Gm10413
Entrez Gene ID
100041774
Gene Name
predicted gene 10413
Gene Aliases
100038846, 1700001E04Rik, ENSMUSG00000072749, Gm1973
Chromosome Location
Chr. 14: 3395093 - 3403534 on Build GRCm38
UniGene ID
Mm.360540
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029288.3 NP_083564.2 1 178
GenBank mRNA AK005583.1 - 1 168
BC100412.1 - 1 178 30916569

View Full Product Details